Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: skn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380237.1 | NP_001367379.1 | 2 - 3 | 750 | 71 | |
NM_171345.6 | NP_741404.1 | 4 - 5 | 1020 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |