Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306912.2 | NP_001293841.1 | 5 - 6 | 1087 | 69 | |
NM_001306913.3 | NP_001293842.1 | 6 - 7 | 1153 | 69 | ||
NM_001306914.2 | NP_001293843.1 | 5 - 6 | 1051 | 69 | ||
NM_001380288.1 | NP_001368430.1 | 4 - 5 | 852 | 69 | ||
NM_001383109.1 | 6 - 7 | 1110 | 69 | |||
NM_171367.8 | NP_741430.3 | 5 - 6 | 984 | 69 | ||
GenBank mRNA | AY455928.1 | - | 5 - 6 | 1046 | 69 | |
AY493413.1 | - | 5 - 6 | 1038 | 69 | ||
AY493414.1 | - | 4 - 5 | 867 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |