Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jnk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026099.5 | NP_001021270.1 | 1 - 2 | 84 | 80 | |
NM_001383114.1 | 1 - 2 | 90 | 80 | |||
NR_131561.1 | 1 - 2 | 48 | 80 | |||
GenBank mRNA | AB024085.1 | - | 1 - 2 | 91 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |