Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171372.3 | NP_741435.2 | 5 - 6 | 765 | 124 | |
NM_171373.6 | NP_741436.2 | 3 - 4 | 423 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |