Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171372.3 | NP_741435.2 | 6 - 7 | 1301 | 162 | |
NM_171373.6 | NP_741436.2 | 4 - 5 | 959 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |