Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: deb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306814.4 | NP_001293743.1 | 6 - 7 | 1432 | 70 | |
NM_001306815.3 | NP_001293744.1 | 6 - 7 | 1432 | 70 | ||
NM_068703.7 | NP_501104.2 | 5 - 6 | 1192 | 70 | ||
NM_171374.6 | NP_741437.1 | 5 - 6 | 1192 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |