Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027998.6 | NP_001023169.1 | 3 - 4 | 622 | 74 | |
NM_171382.4 | NP_741445.1 | 3 - 4 | 635 | 74 | ||
GenBank mRNA | AF083226.1 | - | 3 - 4 | 623 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |