Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383183.1 | 9 - 10 | 1140 | 66 | ||
NM_059334.5 | NP_491735.1 | 7 - 8 | 1023 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |