Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps-34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380450.1 | NP_001367304.1 | 1 - 2 | 154 | 110 | |
NM_001383194.1 | 1 - 2 | 170 | 110 | |||
NM_059340.5 | NP_491741.1 | 1 - 2 | 166 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |