Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: haf-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059353.7 | NP_491754.1 | 4 - 5 | 675 | 112 | |
NM_182010.5 | NP_871810.1 | 4 - 5 | 658 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |