Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: haf-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059353.7 | NP_491754.1 | 9 - 10 | 1239 | 81 | |
NM_182010.5 | NP_871810.1 | 9 - 10 | 1222 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |