Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cof-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059403.5 | NP_491804.2 | 7 - 8 | 1136 | 114 | |
GenBank mRNA | AY494975.1 | - | 7 - 8 | 1040 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |