Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bli-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026369.6 | NP_001021540.1 | 9 - 10 | 1683 | 106 | |
NM_001026372.7 | NP_001021543.1 | 9 - 10 | 1689 | 106 | ||
NM_001316849.3 | NP_001303778.1 | 10 | 1689 | 106 | ||
NM_001316850.3 | NP_001303779.1 | 9 - 10 | 1685 | 106 | ||
NM_001316852.3 | NP_001303781.1 | 9 - 10 | 1685 | 106 | ||
NM_001372959.2 | NP_001360008.1 | 10 | 1690 | 106 | ||
NM_001380517.2 | NP_001367786.1 | 9 - 10 | 1671 | 106 | ||
NM_001380518.2 | NP_001367785.1 | 10 - 11 | 1733 | 106 | ||
NM_001380521.1 | NP_001368583.1 | 8 - 9 | 1525 | 106 | ||
NM_001380523.1 | NP_001367787.1 | 8 - 9 | 1525 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |