Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001361754.3 | NP_001348668.1 | 7 - 8 | 1669 | 69 | |
NM_059465.5 | NP_491866.2 | 7 - 8 | 1670 | 69 | ||
GenBank mRNA | AY887907.1 | - | 7 - 8 | 1686 | 69 | |
AY954523.1 | - | 7 - 8 | 1686 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |