Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mdt-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383258.1 | 2 - 3 | 127 | 121 | ||
NM_059468.6 | NP_491869.1 | 2 - 3 | 162 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |