Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smg-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059528.8 | NP_491929.1 | 4 - 5 | 965 | 103 | |
GenBank mRNA | U64441.1 | - | 3 - 4 | 796 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |