Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025858.4 | NP_001021029.1 | 4 - 5 | 1042 | 112 | |
NM_001306351.3 | NP_001293280.1 | 2 - 3 | 380 | 112 | ||
NM_059539.6 | NP_491940.2 | 4 - 5 | 801 | 112 | ||
GenBank mRNA | AF170425.1 | - | 4 - 5 | 811 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |