Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059544.7 | NP_491945.2 | 9 - 10 | 2888 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |