Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025882.4 | NP_001021053.1 | 3 - 4 | 301 | 125 | |
NM_001374908.2 | NP_001361976.1 | 3 | 273 | 125 | ||
GenBank mRNA | AF149821.1 | - | 3 - 4 | 301 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |