Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276522.3 | NP_001263451.1 | 5 - 6 | 793 | 84 | |
NM_001276523.3 | NP_001263452.1 | 4 - 5 | 646 | 84 | ||
NM_059592.6 | NP_491993.1 | 7 - 8 | 1355 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |