Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sem-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059596.5 | NP_491997.1 | 9 - 10 | 1960 | 60 | |
GenBank mRNA | U60112.1 | - | 9 - 10 | 1960 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |