Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glr-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059616.3 | NP_492017.3 | 1 - 2 | 70 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |