Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rde-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026227.3 | NP_001021398.1 | 2 - 3 | 386 | 169 | |
NM_001026228.4 | NP_001021399.1 | 2 - 3 | 439 | 169 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |