Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inx-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059677.7 | NP_492078.1 | 3 - 4 | 499 | 93 | |
NM_059678.6 | NP_492079.1 | 3 - 4 | 481 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |