Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inx-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059677.7 | NP_492078.1 | 7 - 8 | 1367 | 67 | |
NM_059678.6 | NP_492079.1 | 7 - 8 | 1343 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |