Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136283.3 | NP_001129755.1 | 7 - 8 | 1154 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |