Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059708.4 | NP_492109.2 | 11 - 12 | 1407 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |