Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059712.6 | NP_492113.2 | 1 - 2 | 252 | 112 | |
GenBank mRNA | AY874871.1 | - | 1 - 2 | 215 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |