Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059712.6 | NP_492113.2 | 2 - 3 | 398 | 62 | |
GenBank mRNA | AY874871.1 | - | 2 - 3 | 361 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |