Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rlbp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001263636.3 | NP_001250565.1 | 4 - 5 | 373 | 81 | |
NM_001267076.2 | NP_001254005.1 | 7 - 8 | 1006 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |