Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gna-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059743.7 | NP_492144.1 | 2 - 3 | 283 | 128 | |
GenBank mRNA | AY455927.1 | - | 1 - 2 | 356 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |