Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gna-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059743.7 | NP_492144.1 | 3 - 4 | 454 | 72 | |
GenBank mRNA | AY455927.1 | - | 2 - 3 | 527 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |