Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129010.3 | NP_001122482.2 | 3 - 4 | 438 | 90 | |
NM_001129011.4 | NP_001122483.2 | 3 - 4 | 399 | 90 | ||
GenBank mRNA | EF601077.1 | - | 3 - 4 | 399 | 90 | |
EF601082.1 | - | 3 - 4 | 438 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |