Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ath-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306367.3 | NP_001293296.1 | 4 - 5 | 703 | 91 | |
NM_001383433.1 | 4 - 5 | 729 | 91 | |||
NR_131388.1 | 4 - 5 | 833 | 91 | |||
NR_131389.1 | 3 - 4 | 645 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |