Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026376.5 | NP_001021547.1 | 6 - 7 | 3494 | 114 | |
NM_059816.7 | NP_492217.2 | 6 - 7 | 3494 | 114 | ||
GenBank mRNA | AF013950.1 | - | 6 - 7 | 3488 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |