Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025822.3 | NP_001020993.1 | 1 - 2 | 319 | 82 | |
NM_001025823.6 | NP_001020994.1 | 1 - 2 | 324 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |