Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025825.5 | NP_001020996.1 | 5 - 6 | 755 | 114 | |
GenBank mRNA | AJ133374.1 | - | 5 - 6 | 752 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |