Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhs-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129036.3 | NP_001122508.1 | 3 - 4 | 394 | 96 | |
NM_001129037.4 | NP_001122509.1 | 2 - 3 | 389 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |