Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kri-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026692.5 | NP_001021863.1 | 4 - 5 | 654 | 88 | |
NM_001026693.5 | NP_001021864.1 | 4 - 5 | 649 | 88 | ||
GenBank mRNA | DQ372920.1 | - | 4 - 5 | 649 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |