Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318276.4 | NP_001305205.1 | 3 | 347 | 77 | |
NM_059845.4 | NP_492246.2 | 2 - 3 | 371 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |