Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sre-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059846.3 | NP_492247.2 | 6 - 7 | 755 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |