Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059855.6 | NP_492256.1 | 5 - 6 | 1738 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |