Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ser-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059872.3 | NP_492273.2 | 1 - 2 | 178 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |