Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sod-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059889.6 | NP_492290.1 | 3 - 4 | 273 | 104 | |
GenBank mRNA | D12984.1 | - | 2 - 3 | 242 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |