Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059903.6 | NP_492304.1 | 2 - 3 | 976 | 100 | |
GenBank mRNA | AF286377.1 | - | 2 - 3 | 715 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |