Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gon-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129034.3 | NP_001122506.1 | 13 - 14 | 2861 | 94 | |
NM_001136316.4 | NP_001129788.1 | 7 - 8 | 1826 | 94 | ||
NM_001373178.2 | NP_001359707.1 | 15 - 16 | 3012 | 94 | ||
NM_059914.7 | NP_492315.4 | 16 - 17 | 3085 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |