Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: edc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059927.6 | NP_492328.1 | 7 - 8 | 1077 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |