Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hot-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059948.6 | NP_492349.1 | 2 - 3 | 256 | 107 | |
GenBank mRNA | AF324057.1 | - | 2 - 3 | 129 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |