Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hot-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059948.6 | NP_492349.1 | 3 - 4 | 378 | 97 | |
GenBank mRNA | AF324057.1 | - | 3 - 4 | 251 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |