Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpb-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059960.7 | NP_492361.2 | 4 - 5 | 642 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |